ContactUsLinkComponent
Portable refractometers for special sugars
Portable refractometers for special sugars
Supplier:  Atago
Certificates
undefined
Portable refractometers for special sugars
Supplier:  Atago
Specifications
  • Catalog No:
  • 635-0223
  • 635-0224
  • 635-0222
  • 635-0228
  • 635-0226
  • 635-0227
  • 635-0225
  • 635-0220
  • Response time:
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • Electrical:
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • W×D×H:
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • Weight:
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g