ContactUsLinkComponent
Formaldehyde monitor
Formaldehyde monitor
Catalog # 514-0008
Supplier:  3M
undefined
Formaldehyde monitor
Catalog # 514-0008
Supplier:  3M
Specifications
  • Description:
    Formaldehyde monitors
  • Seq:
    02
  • Pk:
    5
Specifications
About this item
Monitors use the proven scientific principle of diffusion, which involves the gradual mixing of molecules of two or more substances.
  • Formaldehyde monitor uses a bisulphite-impregnated filter and is analysed by colorimetry
  • Can be used for eight-hour TWA sampling
  • Does not have the sensitivity for short-term exposure limit (STEL) monitoring
Designed for measuring personal exposure in work areas in a variety of industries.
18-month shelf life.
Certifications: Meet the OSHA requirement of ±25% accuracy at 0,5 - 5,0 ppm.