Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody (APC-Cyanine7) [clone: GK1.5]
Anti-CD4 Rat Monoclonal Antibody (APC-Cyanine7) [clone: GK1.5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD4 Rat Monoclonal Antibody (APC-Cyanine7) [clone: GK1.5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-0041-U100
  • 25-0041-U025
  • Antigen symbol:
  • L3T4, T7
  • L3T4, T7
  • Antigen name:
  • CD4
  • CD4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • GK1.5
  • GK1.5
  • Reactivity:
  • Syrian hamster, Mouse
  • Syrian hamster, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together