Zadání objednávky
Czech Republic
ContactUsLinkComponent
Nitric oxide assay kit
Nitric oxide assay kit
  ABNOKA1641
 :  Abnova
 :  
undefined
Nitric oxide assay kit
  ABNOKA1641
 :  Abnova
 :  
  • Bal.:
    1 KIT
  • Popis:
    Nitric Oxide Assay Kit, 96 well
Nitric Oxide Assay Kit for the quantitative colorimetric determination of nitric oxide in cell, food sample, plasma, serum, tissue and urine.
  • Detection range: 0,6 to 200 µm
  • Sample volume: 100 µl
 : The kit contains reagent A (12 ml), reagent B (500 µl) and reagent C (12 ml), NaOH (1 ml), ZnSO4 (1 ml) and standard (1 ml).