Human MyD88 ELISA kit
Catalog # BIRBORB106601-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Human
- Description:Human MyD88 ELISA Kit
- Detection range:312 - 20000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Human MyD88 ELISA Kit for the quantitative detection of human Myeloid differentiation primary response gene 88 (MYD88).
MYD88 is a protein that, in humans, is encoded by the MYD88 gene. By interspecific backcross mapping, the mouse MyD88 gene was localised to chromosome 9; the human homolog was mapped to 3p22-p21. The human ortholog MYD88 seems to function similarly to mice, however, available evidence suggests that MYD88 is dispensable for human resistance to common viral infections and to all but a few pyogenic bacterial infections, demonstrating a major difference between mouse and human immune responses.