Square cover glasses for fluorescence
Supplier: MARIENFELD
Certificates
About this item
These cover glasses are made of borosilicate glass D 263™ M of the hydrolytic class 1.
- Colourless and clear
- Chemically resistant
Suitable for fluorescence microscopy.
Certifications: ISO 8255 - Approved for in vitro diagnostic (IVD) applications according to IVD directive 98/79/EC.
Packaging: In hinged lid plastic boxes of 100 or 200 pieces - Ten boxes in a carton.
Frequently Bought Together