Human recombinant INF beta 1a (from CHO cells)
Catalog # BIRBORB80387-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Storage conditions:Lyophilized Interferon-beta 1a although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution IFN-Beta1a should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Biological activity:The specific activity as determined in viral resistance assay (human "Wish" cell line and VSV virus or the monkey VERO cell line with EMCV virus) was found to be 270,000,000 IU/mg.
- Protein synonyms:Leukocyte interferon
- Protein/peptide name:INF beta 1a
- Purity:Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized Interferon beta 1a in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MSYNLLGFLQ RSSNFQCQKL LWQLNGRLEY CLKDRMNFDI PEEIKQLQQF QKEDAALTIY EMLQNIFAIF RQDSSSTGWN ETIVENLLAN VYHQINHLKT VLEEKLEKED FTRGKLMSSL HLKRYYGRIL HYLKAKEYSH CAWTIVRVEI LRNFYFINRL TGYLRN.
- Formulation:Lyophilized from 1mg/10ml solution containing 91 mg Human Albumin, and 620 mg mannitol, and mg Sodium Acetate.
- Tested applications:SDS-PAGE