Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant CETN3 (from E. coli)
Human recombinant CETN3 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant CETN3 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    CETN3 Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze thaw cycles.
  • Protein synonyms:
    CEN3
  • Protein/peptide name:
    CETN3
  • Purity:
    Greater than 95% as determined by SDS-PAGE.
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH MSLALRSELV VDKTKRKKRR ELSEEQKQEI KDAFELFDTD KDEAIDYHEL KVAMRALGFD VKKADVLKILKDYDREATGK ITFEDFNEVV TDWILERDPH EEILKAFKLF DDDDSGKISL RNLRRVAREL GENMSDEELR AMIEEFDKDG DGEINQEEFI AIMTGDI.
  • Formulation:
    CETN3 Human 0.5mg/ml solution contains 20 mM Trsi HCL pH-8, 0.1M NaCl and 10% glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together