Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant CD45 (from E. coli)
Human recombinant CD45 (from E. coli)
  BIRBORB81010-5
 :  Biorbyt
 :  
Human recombinant CD45 (from E. coli)
  BIRBORB81010-5
 :  Biorbyt
 :  

 

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
  • Protein synonyms:
    CD 45
  • Protein/peptide name:
    CD45
  • Formulation:
    CD45 at 100ug/1ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS