Human recombinant U1snRNP-68 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:U1 small nuclear ribonucleoprotein 70 kDa
- Protein/peptide name:U1snRNP-68
- Purity:Greater than 90% as determined by SDS-PAGE.
- Formulation:U1-snRNP is supplied in 20 mM HEPES buffer pH-7.5, 0.01mM EDTA and 0.02% SDS.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together