Order Entry
Czech Republic
ContactUsLinkComponent
 
Pig Porcine Leptin (from E. coli)
 :  Biorbyt
 
Pig Porcine Leptin (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Pig
  • Storage conditions:
    Lyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
  • Protein synonyms:
    LEP
  • Protein/peptide name:
    Porcine Leptin
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together