Human VEGFR2 (from insect cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Insect cells
- Species:Human
- Storage conditions:Lyophilized VEGFR-2 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution FLK1 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:The activity of VEGFR-2 D1-7 was determined by its ability to abolish the binding of iodinated VEGF to solid surfaces or cell surfaces receptors. The ED50 for this effect is typically 10.0 ng/ml.
- Protein synonyms:CD309
- Protein/peptide name:VEGFR2
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized VEGFR2 in sterile water not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:A SVGLPSVSLD LPRLSIQKDI LTIKANTTLQ ITCRGQRDLD WLWPNNQSGS EQRVEVTECS DGLFCKTLTI PKVIGNDTGA YKCFYRETDL ASVIYVYVQD YRSPFIASVS DQHGVVYITE NKNKTVVIPC LGSISNLNVS LCARYPEKRF VPDGNRISWD SKKGFTIPSY MISYAGMVFC EAKINDESYQ SIMYIVVVVG YRIYDVVLSP SHGIELSVGE KLVLNCTART ELNVGIDFNW EYPSSKHQHK KLVNRDLKTQ SGSEMKKFLS TLTIDGVTRS DQGLYTCAAS SGLMTKKNST FVRVHEKPFV AFGSGMESLV EATVGERVRI PAKYLGYPPP EIKWYKNGIP LESNHTIKAG HVLTIMEVSE RDTGNYTVIL TNPISKEKQS HVVSLVVYVP PQIGEKSLIS PVDSYQYGTT QTLTCTVYAI PPPHHIHWYW QLEEECANEP SQAVSVTNPY PCEEWRSVED FQGGNKIEVN KNQFALIEGK NKTVSTLVIQ AANVSALYKC EAVNKVGRGE RVISFHVTRG PEITLQPDMQ PTEQESVSLW CTADRSTFEN LTWYKLGPQP LPIHVGELPT PVCKNLDTLW KLNATMFSNS TNDILIMELK NASLQDQGDY VCLAQDRKTK KRHCVVRQLT VLERVAPTIT GNLENQTTSI GESIEVSCTA SGNPPPQIMW FKDNETLVED SGIVLKDGNR NLTIRRVRKE DEGLYTCQAC SVLGCAKVEA FFIIEGA.
- Formulation:KDR was lyophilized from concentrated (1 mg/ml) sterile solution containing 25mM MES pH-5.5 and 100mM NaCl.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together