Order Entry
Czech Republic
ContactUsLinkComponent
Mouse neurotrophin 3 (from E. coli)
Mouse neurotrophin 3 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse neurotrophin 3 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized NGF2 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution NGF-2 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant induction of choline acetyl transferase in rat basal forebrain primary septal culture was found between 20-50 ng/ml.
  • Protein synonyms:
    Nerve growth factor-2
  • Protein/peptide name:
    Neurotrophin 3
  • Purity:
    Greater than 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Neurotrophin-3 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Tyr-Ala-Glu-His-Lys.
  • Formulation:
    Lyophilized from 0.02% TFA.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together