Human VEGF (121) (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Liquid VEGF 121 although stable at 10 °C for week, should be stored below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:The ED50 for this effect is <1.8ng/ml. Measured in cell proliferation assay using NIH-3T3 cell.
- Protein synonyms:Vascular endothelial growth factor A
- Protein/peptide name:VEGF (121)
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQEKCDKP RR.
- Formulation:VEGF-121 His Tag in 20 mM TRIS pH-8.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together