Rat VEGF-C (152) (from insect cells (Sf9))
Dodavatel: Biorbyt
Certifikáty
O tomto produktu
Specifikace
- Source:Insect cells (Sf9)
- Species:Rat
- Storage conditions:Lyophilized Vascular Endothelial Growth Factor-C152 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution VEGF-C 152 should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Biological activity:Measured by its ability to stimulate phosphorylation of the VEGFR-3/FLT-4 receptor in porcine aortic endothelial cells (PAE/FLT -4C ells). The ED50 for this effect is typically 150-300 ng/ml, corresponding to specific activity of 5000 Units/mg.
- Protein synonyms:Vascular endothelial growth factor 152
- Protein/peptide name:VEGF-C (152)
- Purity:Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Vascular Endothelial Growth Factor 152 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:The protein was lyophilized from concentrated (1mg/ml) solution with BSA.
- Tested applications:SDS-PAGE
Specifikace
Frequently Bought Together