Recombinant GRA7
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Storage conditions:Store at 4 °C. Upon arrival, store at −20 °C
- Protein/peptide name:GRA7
- Purity:Toxoplasma protein is >95.0% pure as determined by 12.0% PAGE
- Formulation:50 mM Tris-HCl and 50% glycerol. No preservative and no stabilizer
- Tested applications:ELISA , WB
Specifications
Frequently Bought Together