Recombinant HPV 18 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Storage conditions:Two years when when kept frozen at -2 °C to −80 °C.
- Protein synonyms:Papillomavirus
- Protein/peptide name:HPV 18
- Purity:Greater than 95% pure as determined by 10% PAGE (Coomassie staining). The recombinant HPV-16 fusion protein was purified by GSH affinity chromatography technique.
- Formulation:The protein is formulated with 1x PBS and 0.5mM EDTA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together