Order Entry
Czech Republic
ContactUsLinkComponent
HIV-1 24 HRP (from E. coli)
HIV-1 24 HRP (from E. coli)
Supplier:  Biorbyt
Certificates
HIV-1 24 HRP (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    HIV-1 p24 protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HIV-1 24 HRP
  • Purity:
    Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
  • Formulation:
    50mM Tris pH-7.2 8M urea.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together