Order Entry
Czech Republic
ContactUsLinkComponent
Human MOG (from E. coli)
Human MOG (from E. coli)
Supplier:  Biorbyt
Certificates
Human MOG (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized MOG although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution MOG should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Myelin Oligodendrocyte Glycoprotein
  • Protein/peptide name:
    MOG
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized MOG in sterile 10mM Acetic acid not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    The Myelin Oligodendrocyte Glycoprotein 0.5mg/ml solution was lyophilized from 20 mM sodium acetate buffer pH-4 and 0.3M sodium chloride.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together