TmpA partial
Catalog # BIRBORB98097-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Storage conditions:Store at−20 °C.
- Protein/peptide name:TmpA partial
- Purity:Greater than 90% by SDS-PAGE
- Formulation:Liquid. Supplied in 10mM Tris-HCl pH8.0, 20mM DDT,1mM EDTA and 8M urea.
- Tested applications:ELISA , WB