HBV-Core 16 kDa
100 µG
BIRBORB97877-100
Marketsource
: Biorbyt
- Pk:100 µG
- Cat. No.:BIRBORB97877-100
- Protein/peptide name:HBV-Core 16 kDa
- Storage conditions:Store at−20 °C.
- Formulation:Liquid. Supplied as a 1 mg/ml solution in 10 mM Tris-HClpH 8.0, 50 mM NaCl, 1 mM EDTA and 50% glycerol.
- Purity:Greater than 95% by SDS-PAGE (coomassie staining)and RP-HPLC.
- Tested applications:ELISA



