Order Entry
Czech Republic
ContactUsLinkComponent
HBV-Core 18 kDa
HBV-Core 18 kDa
Catalog # BIRBORB97876-100
Supplier:  Biorbyt
CAS Number:  
HBV-Core 18 kDa
Catalog # BIRBORB97876-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    HBV-Core 18 kDa
  • Purity:
    Greater than 95% by SDS-PAGE (coomassie staining)and RP-HPLC.
  • Formulation:
    Liquid. Supplied as a 1 mg/ml solution in 10 mM Tris-HClpH 8.0, 50 mM NaCl, 1 mM EDTA and 50% glycerol.
  • Tested applications:
    ELISA

Specifications

About this item