Human recombinant IL1RA (fromE. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Biological activity:ED50 is 0.5 ng/ml as determined by the dosedependent inhibition of IL1 stimulation of D10S cells.
Specific Activity of 2.0 x 106 IU/mg. - Protein synonyms:DIRA
- UniProtKB:P18510 (Human)
- Protein/peptide name:IL1RA
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MRPSGRKSSKMQAFRIWDVNQKTFYLRNNQLVAGYLQGPNVNLEEKIDVVPIEPHALFLGIHGGK MCLSCVKSGDETRLQLEAVNITDLSENRKQDKRFAFIRSDSGPTTSFESAACPGWFLCTAMEADQ PVSLTNMPDEGVMVTKFYFQEDE
- Formulation:Lyophilized from a 0.2 um filtered solution of 50mM TrisHCl, 200mM NaCl, pH 7.5.
- Tested applications:SDS-PAGE , FuncS
Specifications
Frequently Bought Together