Human recombinant CNDP1 (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:His dipeptide (carnosine)
- UniProtKB:Q96KN2 (Human)
- Protein/peptide name:CNDP1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:PSPPPALLEKVFQYIDLHQDEFVQTLKEWVAIESDSVQPVPRFRQELFRMMAVAADTLQRLGARV ASVDMGPQQLPDGQSLPIPPVILAELGSDPTKGTVCFYGHLDVQPADRGDGWLTDPYVLTEVDGK LYGRGATDNKGPVLAWINAVSAFRALEQDLPVNIKFIIEGMEEAGSVALEELVEKEKDRFFSGVD YIVISDNLWISQRKPAITYGTRGNSYFMVEVKCRDQDFHSGTFGGILHEPMADLVALLGSLVDSS GHILVPGIYDEVVPLTEEEINTYKAIHLDLEEYRNSSRVEKFLFDTKEEILMHLWRYPSLSIHGI EGAFDEPGTKTVIPGRVIGKFSIRLVPHMNVSAVEKQVTRHLEDVFSKRNSSNKMVVSMTLGLHP WIANIDDTQYLAAKRAIRTVFGTEPDMIRDGSTIPIAKMFQEIVHKSVVLIPLGAVDDGEHSQNE KINRWNYIEGTKLFAAFFLEMAQLHVDHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 7.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together