Recombinant FGF1
Catalog # BIRBORB85465-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 10 ng/ml corresponding to a specific activity of 105 Units/mg, calculated by the dose dependentproliferation of BAF3 cells expressing FGF receptorsmeasured by 3H-thymidine uptake.
- UniProtKB:P05230
- Protein/peptide name:FGF1
- Purity:Greater than 95% by SDS-PAGE and RP-HPLC
- Formulation:Lyophilized. Recombinant FGF-1 was lyophilized from a1 mg/ml solution containing 10 mM sodium phosphatepH 7.5 and 75 mM NaCl.
- Tested applications:WB , SDS-PAGE