Hamster Apolipoprotein AI (from Plasma)
Catalog # BIRBORB81949-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Source:Plasma
- Species:Hamster
- Protein synonyms:APOA1
- Protein/peptide name:Apolipoprotein AI
- Purity:Purified.
- Formulation:Preservative: 0.05% Thimerosal
- Purification:Purified