Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Neurotrophin 3 (from E. coli)
Human recombinant Neurotrophin 3 (from E. coli)
Catalog # BIRBORB82075-10
Supplier:  Biorbyt
CAS Number:  
Human recombinant Neurotrophin 3 (from E. coli)
Catalog # BIRBORB82075-10
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    Neurotrophin 3 ELISA Kit
  • Protein/peptide name:
    Neurotrophin 3
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.

Specifications

About this item