Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant Leptin (from E. coli)
  BIRBORB82068-1
 :  Biorbyt
 :  
Human recombinant Leptin (from E. coli)
  BIRBORB82068-1
 :  Biorbyt
 :  

 

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    LEP
  • Protein/peptide name:
    Leptin
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.