Human recombinant C-Reactive Protein (from E. coli)
Catalog # BIRBORB81870-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:CRP
- Protein/peptide name:C-Reactive Protein
- Purity:Purified.
- Formulation:Preservative: 0.05% Sodium Azide