Order Entry
Czech Republic
ContactUsLinkComponent
Human s-100b, protein (beta-beta) (from brain)
Human s-100b, protein (beta-beta) (from brain)
Catalog # BIRBORB82231-1
Supplier:  Biorbyt
CAS Number:  
Human s-100b, protein (beta-beta) (from brain)
Catalog # BIRBORB82231-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Brain
  • Species:
    Human
  • Protein synonyms:
    S-1beta
  • Protein/peptide name:
    S-100B, Protein (beta-beta)
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.
  • Tested applications:
    ELISA

Specifications

About this item