Rubella recombinant rubella virus E2 protein (from E. coli)
Catalog # BIRBORB82463-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rubella
- Protein synonyms:Rubella Virus E2
- Protein/peptide name:Rubella Virus E2 protein
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB