Rubella rubella grade IV antigen (Strain HPV77) >90% viral protein (from vero cells)
Catalog # BIRBORB81835-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:Vero cells
- Species:Rubella
- Protein synonyms:Rubella Grade IV Antigen (Strain HPV77) >90% Viral
- Protein/peptide name:Rubella Grade IV Antigen (Strain HPV77) >90% Viral Protein
- Purity:>90%
- Formulation:Not lyophilised.
- Tested applications:ELISA
- Purification:Purified