Human TNF alpha (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:APC1
- Protein/peptide name:TNF alpha
- Purity:> 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:TNF-a His is supplied in 1xPBS, 50% glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together