Human MIF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:Fully biologically active measured by its ability to bind rhCD74 in a functional ELISA.
- Protein synonyms:acrophage migration inhibitory factor (glycosylation-inhibiting factor)
- Protein/peptide name:MIF
- Purity:> 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized MIF in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MPMFIVNTNV PRASVPDGFL SELTQQLAQA TGKPPQYIAV HVVPDQLMAF GGSSEPALCS LHSIGKIGGA QNRSYSKLLC GLLAERLRIS PDRVYINYYD MNAANVGWNN STFALEHHHH HH.
- Formulation:Human MIF was lyophilized from 1mg/ml solution containing PBS pH-7.4.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together