Order Entry
Czech Republic
ContactUsLinkComponent
Mouse IL13 (from E. coli)
Mouse IL13 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse IL13 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 range=2-6 ng/ml, determined by the dose dependent prolifiration of TF-1 cells.
  • Protein synonyms:
    Allergic rhinitis
  • Protein/peptide name:
    IL13
  • Purity:
    > 95% as determined by SDS-PAGE.
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin 13 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and found to be Met-Pro-Val-Pro-Arg.
  • Formulation:
    The protein (1mg/ml) was lyophilized with no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together