Order Entry
Czech Republic
ContactUsLinkComponent
Bovine Recombinant bPI3Ka (from Insect cells (Sf9))
Bovine Recombinant bPI3Ka (from Insect cells (Sf9))
Supplier:  Biorbyt
Certificates
Bovine Recombinant bPI3Ka (from Insect cells (Sf9))
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Bovine
  • Storage conditions:
    PI3Ka although stable at 14 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Please prevent freeze-thaw cycles.
  • Biological activity:
    1,000 U/mg.
  • Protein synonyms:
    Phosphoinositide 3-kinase alpha p110a
  • Protein/peptide name:
    bPI3Ka
  • Purity:
    Greater than 90.0% as determined by SDS-PAGE.
  • Formulation:
    PI3Ka solution in 25mM HEPES, pH 8.0, 25mM NaCl, 2.5mM MgCl2 and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together