Mouse recombinant G-CSF
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Storage conditions:Store at −20 °C.
- Biological activity:The ED50 was determined by the dose-dependent stimulation of the proliferation of Mouse NFS-60 cells is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 107 units/mg.
- Protein synonyms:Granulocyte Colony-Stimulating Factor
- UniProtKB:P09920
- Protein/peptide name:G-CSF
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Granulocyte Colony Stimulating Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
- Formulation:GCSF was lyophilized with no additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together