Order Entry
Czech Republic
ContactUsLinkComponent
Adenosine-5’-Triphosphate (ATP) 100 mM in aqueous solution ≥99% for molecular biology
Adenosine-5’-Triphosphate (ATP) 100 mM in aqueous solution ≥99% for molecular biology
Catalog # R0441
CAS Number:  
Adenosine-5’-Triphosphate (ATP) 100 mM in aqueous solution ≥99% for molecular biology
Catalog # R0441
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    0.25 ml

Specifications

About this item

 :
 :595.11 g/mol
Formula:C₁₀H₁₂N₅O₁₃P₃Na₄