ContactUsLinkComponent
Beakers, low form, Corning®
Beakers, low form, Corning®
Catalog # CORN1000P-150
Supplier:  Corning
Supplier #: 1000P-150
undefined
Beakers, low form, Corning®
Catalog # CORN1000P-150
Supplier:  Corning
Supplier #: 1000P-150
Log in for availability
Select an Option
56 mm
Select an Option
20 ml
Select an Option
150 ml
Select an Option
80 mm
Select an Option
04
Select an Option
12
About this item
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
With printed blue graduations.
Frequently Bought Together