ContactUsLinkComponent
One-piece fittings, Super Flangeless™, Upchurch Scientific®
One-piece fittings, Super Flangeless™, Upchurch Scientific®
Catalog # UPCHP-249
undefined
One-piece fittings, Super Flangeless™, Upchurch Scientific®
Catalog # UPCHP-249
Specifications
  • Description:
    P-249 One-piece super flangeless fitting, 1/4-28 flat bottom port, standard knurl head style
  • Material:
    PEEK
  • Pk:
    1
  • For tubing ext.Ø:
    1/16"
Specifications
About this item
One piece super flangeless fitting is ideal for those connections that need to be broken frequently and tubing assemblies because they swage onto the tubing.
  • Excellent chemical resistance