Order Entry
Czech Republic
ContactUsLinkComponent
Leak diverter bucket kit with absorbents, PIG® Roof Leak Diverter
Leak diverter bucket kit with absorbents, PIG® Roof Leak Diverter
Catalog # NEWPTLS667-TR
Supplier:  NEW PIG
undefined
Leak diverter bucket kit with absorbents, PIG® Roof Leak Diverter
Catalog # NEWPTLS667-TR
Supplier:  NEW PIG

Specifications

  • Description:
    Leak diverter kit, 5,4 kg
  • L×W:
    1524×1524 mm
  • Pk:
    1 KIT

Specifications

About this item

This leak diverter kit features a pail lid with a removable plug to allow the drainage hose to be inserted without having to remove the lid. This can be used as a collection container for diverted liquid.

  • Non-permeable tarp is vinyl coated for strength and durability
  • Resistance to punctures, UV and mildew for long life
  • Flow rate is 12,6 litres/minute when used with a 12 mm ID clear drainage hose
  • Adjustable straps allow fast setup and response
  • Perfect for rapid response to overhead leaks
Ordering information: Includes a drainage hose (635 mm), 4× adjustable hanging straps (1524 mm), a bucket (246 litres), a 'CAUTION' absorbent mat pads (30 packs) and two all purpose wipers (25 packs).