ContactUsLinkComponent
Vacuum manifold
Vacuum manifold
Catalog # ENZOADI987500
Supplier:  ENZO LIFE SCIENCES
Vacuum manifold
Catalog # ENZOADI987500
Supplier:  ENZO LIFE SCIENCES
Specifications
  • Description:
    Vacuum manifold
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
The manifold is constructed of a chemical-resistant polymer and is designed to facilitate the filtration of liquid through a multi-well filter plate.