Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 0.0714 mol/l (0.1N/1.4), AVS TITRINORM volumetric solution
Hydrochloric acid 0.0714 mol/l (0.1N/1.4), AVS TITRINORM volumetric solution
Catalog # 307902.400
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid 0.0714 mol/l (0.1N/1.4), AVS TITRINORM volumetric solution
Catalog # 307902.400
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0,07126 - 0,07154 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:0
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324