Hydrochloric acid 0.714 mol/l (N/1.4), AVS TITRINORM volumetric solution
Catalog # 307904.400
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Bag-in-box
- Pk:10 L
- Seq:5214
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0,7126 - 0,7154 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:0
ADR:8,III
:36.46 g/mol
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324