Order Entry
Czech Republic
ContactUsLinkComponent
Anti-BCL2 Mouse Monoclonal Antibody (CF405S) [clone: 100/D5 124]
Anti-BCL2 Mouse Monoclonal Antibody (CF405S) [clone: 100/D5 124]
Supplier:  Biotium
Certificates
Anti-BCL2 Mouse Monoclonal Antibody (CF405S) [clone: 100/D5 124]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041234-100
  • BNC041234-500
  • Antigen symbol:
  • BCL2
  • BCL2
  • Antigen name:
  • bcl-2
  • bcl-2
  • Antigen synonyms:
  • Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
  • Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone 100/D5 124
  • Clone 100/D5 124
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 150749
  • 150749
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together