Anti-KRT7 Mouse Monoclonal Antibody [clone: KRT7/903]
Catalog # ABNOMAB14312
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:KRT8 Recombinant
- Antigen symbol:KRT7
- Clonality:Monoclonal
- Clone:KRT7/903
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:3855
- Amino acid number:P08729
- Storage buffer:In 10 mM PBS.
- Immunogen:Recombinant protein corresponding to full length human KRT7.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human KRT7.
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/903
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human