Anti-TIMM23 Rabbit Polyclonal Antibody
ABGEAP14103B
: Abgent
- Antibody type:Primary
- Antigen name:Translocase of Inner Mitochondrial Membrane 23
- Antigen symbol:TIMM23
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:400 µl
Type: Primary
Antigen: TIMM23
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human