Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KRT7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRT7/760 OV-TL12/30]
Anti-KRT7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRT7/760 OV-TL12/30]
Catalog # BNUM1167-50
Supplier:  Biotium
Anti-KRT7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRT7/760 OV-TL12/30]
Catalog # BNUM1167-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 7
  • Antigen symbol:
    KRT7
  • Clonality:
    Monoclonal
  • Clone:
    Clone KRT7/760 OV-TL12/30
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgGs
  • Reactivity:
    Human,
    Rat
  • Gene ID:
    411501
  • Antigen synonyms:
    KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/760 OV-TL12/30
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Rat