Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KRT7 Mouse Monoclonal Antibody [clone: KRT7/760 OV-TL12/30]
Anti-KRT7 Mouse Monoclonal Antibody [clone: KRT7/760 OV-TL12/30]
Supplier:  Biotium
Certificates
Anti-KRT7 Mouse Monoclonal Antibody [clone: KRT7/760 OV-TL12/30]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB1167-100
  • BNUB1167-500
  • Antigen symbol:
  • KRT7
  • KRT7
  • Antigen name:
  • Cytokeratin 7
  • Cytokeratin 7
  • Antigen synonyms:
  • KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
  • KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone KRT7/760 OV-TL12/30
  • Clone KRT7/760 OV-TL12/30
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 611376
  • 611376
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together