About this item
MAb IPO-10 defines the antigen, which appears on B cell progenitors following HLA-DR and preceding CD10, CD19, CD22, CD37 and cym. It is expressed on resting B cells and than reappears and persists in cytoplasm and on cell surface until cytoplasmic Ig appears. It is a useful antibody for diagnostics of neoplasms of B cell origins. It reacts with human B cell lines Daudi, Raji, Namalva, EB-3, RPMI-8226 (50% of cells). The MAb does not label T cell lines, blood granulocytes, thymocytes or bone marrow stromal fibroblasts. No significant changes are detected after PHA or ConA stimulation while LPS and PWM stimulated cultures after 18-48h show decreased number of antigen-positive cells but in final terms of cultivation antigen is expressed again. This MAb labels B cell leukemias and some lymphomas. Hairy cell leukemia strongly reacts and 70% of B cell CLL and some B-NHL were also positive. IPO-10 reacts with AMML cells and in a majority of Hodgkin's disease cases a significant percentage of affected lymph node cells were detected.
Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: IPO-10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human, Monkey
Specifications
- Catalog No:
- BNCB1090-500
- BNCB1090-100
- Antigen symbol:
- MHC2
- MHC2
- Antigen name:
- MHC II
- MHC II
- Antigen synonyms:
- HLA-DRA
- HLA-DRA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone IPO-10
- Clone IPO-10
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 520048
- 520048
- Isotype:
- IgG3 kappa
- IgG3 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide